Search
Search

TMPRSS9| Gene Symbol: TMPRSS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122425.2 | XP_014977911.2 | 901 | 104 | ||
| XM_028839869.1 | XP_028695702.1 | 943 | 104 | |||
| GenBank mRNA | JV650487.1 | - | 365 | 104 | ||
Set Membership: |
|