Search
Search

RRBP1| Gene Symbol: RRBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828162.1 | XP_028683995.1 | 2416 | 89 | ||
| XM_028828163.1 | XP_028683996.1 | 2407 | 89 | |||
| XM_028828164.1 | XP_028683997.1 | 2386 | 89 | |||
| XM_028828165.1 | XP_028683998.1 | 2416 | 89 | |||
| GenBank mRNA | JU346873.1 | - | 523 | 89 | ||
Set Membership: |
|