Search
Search

ZNF527| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123614.2 | XP_014979100.1 | 249 | 109 | ||
| XM_015123615.2 | XP_014979101.1 | 203 | 109 | |||
| GenBank mRNA | JV073488.1 | - | 119 | 109 | ||
| JV725211.1 | - | 97 | 109 | |||
Set Membership: |
|