Search
Search

TENM4| Gene Symbol: TENM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115338.2 | XP_014970824.2 | 13344 | 99 | ||
| XM_015115340.2 | XP_014970826.2 | 13467 | 99 | |||
| XM_015115341.2 | XP_014970827.2 | 13245 | 99 | |||
| XM_028833869.1 | XP_028689702.1 | 13554 | 99 | |||
| XM_028833870.1 | XP_028689703.1 | 13655 | 99 | |||
| XM_028833871.1 | XP_028689704.1 | 13533 | 99 | |||
| XM_028833872.1 | XP_028689705.1 | 13506 | 99 | |||
| XM_028833873.1 | XP_028689706.1 | 13651 | 99 | |||
| XM_028833874.1 | XP_028689707.1 | 13455 | 99 | |||
| GenBank mRNA | JV577827.1 | - | 111 | 97 | ||
Set Membership: |
|