Search
Search

PLXDC1| Gene Symbol: PLXDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119507.2 | XP_014974993.2 | 1639 | 114 | ||
| XM_028836436.1 | XP_028692269.1 | 2019 | 114 | |||
| XM_028836437.1 | XP_028692270.1 | 1473 | 114 | |||
| GenBank mRNA | JU322964.1 | - | 1425 | 114 | ||
| JV475381.1 | - | 509 | 114 | |||
Set Membership: |
|