Search
Search

WDR36| Gene Symbol: WDR36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140560.2 | XP_014996046.2 | 1188 | 111 | ||
| GenBank mRNA | JU327733.1 | - | 465 | 111 | ||
| JU327734.1 | - | 465 | 111 | |||
| JV052035.1 | - | 1171 | 111 | |||
| JV707144.1 | - | 1166 | 111 | |||
Set Membership: |
|