Search
Search

WASHC5| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145973.2 | XP_015001459.1 | 3115 | 94 | ||
| XM_028852905.1 | XP_028708738.1 | 2920 | 94 | |||
| GenBank mRNA | JU337373.1 | - | 3054 | 94 | ||
| JU478071.1 | - | 3063 | 94 | |||
Set Membership: |
|