Search
Search

LRCH4| Gene Symbol: LRCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134466.2 | XP_014989952.1 | 1297 | 119 | ||
| GenBank mRNA | JU322919.1 | - | 1122 | 119 | ||
| JU475588.1 | - | 1122 | 119 | |||
| JV651411.1 | - | 1113 | 119 | |||
Set Membership: |
|