Search
Search

SLC43A3| Gene Symbol: SLC43A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102854.4 | XP_001102854.1 | 872 | 89 | ||
| XM_015114161.2 | XP_014969647.1 | 837 | 89 | |||
| GenBank mRNA | JU471915.1 | - | 815 | 89 | ||
| JV045579.1 | - | 821 | 89 | |||
Set Membership: |
|