Search
Search

TBC1D8B| Gene Symbol: TBC1D8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127958.2 | XP_014983444.1 | 133 | 106 | ||
| XM_015127959.2 | XP_014983445.1 | 133 | 106 | |||
| XM_028842456.1 | XP_028698289.1 | 133 | 106 | |||
| XM_028842457.1 | XP_028698290.1 | 133 | 106 | |||
| GenBank mRNA | JV585568.1 | - | 116 | 106 | ||
| JV717680.1 | - | 110 | 106 | |||
Set Membership: |
|