Search
Search

TUBGCP3| Gene Symbol: TUBGCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837343.1 | XP_028693176.1 | 2374 | 104 | ||
| XM_028837344.1 | XP_028693177.1 | 2374 | 104 | |||
| XM_028837345.1 | XP_028693178.1 | 2374 | 104 | |||
| GenBank mRNA | JU321752.1 | - | 2243 | 104 | ||
| JU473978.1 | - | 2324 | 104 | |||
| JV045904.1 | - | 2315 | 104 | |||
Set Membership: |
|