Search
Search

DTNBP1| Gene Symbol: DTNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135593.2 | XP_014991079.1 | 689 | 102 | ||
| GenBank mRNA | JU336655.1 | - | 518 | 102 | ||
| JU494657.1 | - | 481 | 102 | |||
| JV048412.1 | - | 593 | 102 | |||
| JV630585.1 | - | 687 | 102 | |||