Search
Search

TERB1| Gene Symbol: TERB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126338.2 | XP_014981824.2 | 724 | 118 | ||
| XM_015126339.2 | XP_014981825.2 | 658 | 118 | |||
| XM_015126341.2 | XP_014981827.2 | 582 | 118 | |||
| XM_015126343.2 | XP_014981829.2 | 726 | 118 | |||
| XM_015126344.2 | XP_014981830.2 | 595 | 118 | |||
| XM_015126346.2 | XP_014981832.2 | 723 | 118 | |||
| XM_015126348.2 | XP_014981834.2 | 722 | 118 | |||
| XM_028841219.1 | XP_028697052.1 | 404 | 118 | |||
| XM_028841220.1 | XP_028697053.1 | 655 | 118 | |||
| XM_028841221.1 | XP_028697054.1 | 328 | 118 | |||
| XM_028841222.1 | XP_028697055.1 | 613 | 118 | |||
| XM_028841223.1 | XP_028697056.1 | 518 | 118 | |||
| XM_028841224.1 | XP_028697057.1 | 547 | 118 | |||
| XM_028841225.1 | XP_028697058.1 | 723 | 118 | |||
| XM_028841226.1 | XP_028697059.1 | 511 | 118 | |||
| GenBank mRNA | JU484762.1 | - | 282 | 118 | ||