Search
Search

LRCH1| Gene Symbol: LRCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121049.2 | XP_014976535.1 | 842 | 66 | ||
| XM_015121050.2 | XP_014976536.1 | 842 | 66 | |||
| XM_015121051.2 | XP_014976537.1 | 842 | 66 | |||
| XM_015121052.2 | XP_014976538.1 | 852 | 66 | |||
| XM_028837200.1 | XP_028693033.1 | 842 | 66 | |||
| GenBank mRNA | JU327189.1 | - | 384 | 66 | ||
| JU327380.1 | - | 384 | 66 | |||
| JV055692.1 | - | 501 | 66 | |||
| JV636050.1 | - | 631 | 66 | |||
Set Membership: |
|