Search
Search

BRWD3| Gene Symbol: BRWD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127728.2 | XP_014983214.1 | 2838 | 72 | ||
| XM_015127729.2 | XP_014983215.1 | 2838 | 72 | |||
| GenBank mRNA | JU331068.1 | - | 2508 | 72 | ||
| JV636076.1 | - | 2595 | 72 | |||