Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07221125_g1
See other ADGRL2 GE Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
LPHN2
Chromosome Location:
Chr.1: 142429463 - 143111519 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07221125_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRL2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015143262.2
XP_014998748.2
2973
88
XM_015143264.2
XP_014998750.2
2353
88
XM_015143272.2
XP_014998758.2
2385
88
XM_015143279.2
XP_014998765.2
2970
88
XM_015143285.2
XP_014998771.2
2958
88
XM_015143290.2
XP_014998776.2
2338
88
XM_015143297.2
XP_014998783.2
2973
88
XM_015143308.2
XP_014998794.2
2971
88
XM_015143318.2
XP_014998804.2
2934
88
XM_015143325.2
XP_014998811.2
2972
88
XM_015143330.2
XP_014998816.2
2934
88
XM_015143342.2
XP_014998828.2
2931
88
XM_015143348.2
XP_014998834.2
2972
88
XM_015143354.2
XP_014998840.2
2969
88
XM_015143359.2
XP_014998845.2
2957
88
XM_015143365.2
XP_014998851.2
2917
88
XM_015143369.2
XP_014998855.2
2930
88
XM_015143370.2
XP_014998856.2
2972
88
XM_015143371.2
XP_014998857.2
2970
88
XM_015143385.2
XP_014998871.2
2930
88
XM_015143389.2
XP_014998875.2
2971
88
XM_015143397.2
XP_014998883.2
2968
88
XM_015143404.2
XP_014998890.2
2932
88
XM_015143411.2
XP_014998897.2
2929
88
XM_015143418.2
XP_014998904.2
2916
88
XM_015143426.2
XP_014998912.2
2312
88
XM_015143428.2
XP_014998914.2
2979
88
XM_015143437.2
XP_014998923.2
2977
88
XM_015143444.2
XP_014998930.2
2977
88
XM_015143451.2
XP_014998937.2
2977
88
XM_015143466.2
XP_014998952.2
2976
88
XM_015143470.2
XP_014998956.2
2976
88
XM_015143471.2
XP_014998957.2
2975
88
XM_015143475.2
XP_014998961.2
2975
88
XM_015143479.2
XP_014998965.2
2975
88
XM_015143487.2
XP_014998973.2
2920
88
XM_015143488.2
XP_014998974.2
2312
88
XM_028832010.1
XP_028687843.1
2421
88
XM_028832032.1
XP_028687865.1
2970
88
XM_028832044.1
XP_028687877.1
2312
88
XM_028832093.1
XP_028687926.1
2956
88
XM_028832123.1
XP_028687956.1
2977
88
XM_028832124.1
XP_028687957.1
2974
88
XM_028832125.1
XP_028687958.1
2972
88
XM_028832129.1
XP_028687962.1
2977
88
XM_028832131.1
XP_028687964.1
2960
88
XM_028832132.1
XP_028687965.1
2920
88
XM_028832133.1
XP_028687966.1
2977
88
XM_028832134.1
XP_028687967.1
2338
88
GenBank mRNA
JU333621.1
-
2201
88
JV043500.1
-
2167
88
JV710532.1
-
2230
88
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
G-protein coupled receptor
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline