Search
Search

HIP1R| Gene Symbol: HIP1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798818.3 | XP_002798864.3 | 1981 | 95 | ||
| XM_015153044.2 | XP_015008530.2 | 2014 | 95 | |||
| XM_028830049.1 | XP_028685882.1 | 1960 | 95 | |||
| XM_028830050.1 | XP_028685883.1 | 1981 | 95 | |||
| GenBank mRNA | JU336992.1 | - | 1873 | 95 | ||
| JU477715.1 | - | 1878 | 95 | |||
| JV632761.1 | - | 1877 | 95 | |||
Set Membership: |
|