Search
Search

HSPH1| Gene Symbol: HSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120828.2 | XP_014976314.2 | 2036 | 70 | ||
| XM_015120829.2 | XP_014976315.2 | 1674 | 70 | |||
| GenBank mRNA | JU332613.1 | - | 1634 | 70 | ||
| JU474076.1 | - | 1638 | 70 | |||
| JV044524.1 | - | 1640 | 70 | |||
| JV492587.1 | - | 1442 | 70 | |||
Set Membership: |
|