Search
Search

NOP14| Gene Symbol: NOP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137885.2 | XP_014993371.2 | 1612 | 80 | ||
| XM_015137886.2 | XP_014993372.2 | 1607 | 80 | |||
| GenBank mRNA | JU336868.1 | - | 1469 | 80 | ||
| JU471619.1 | - | 1488 | 80 | |||
| JV044737.1 | - | 1481 | 80 | |||
| JV489141.1 | - | 1430 | 80 | |||
| JV596930.1 | - | 1480 | 80 | |||
Set Membership: |
|