Search
Search

PPP4R3A| Gene Symbol: PPP4R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144381.2 | XP_014999867.1 | 1715 | 96 | ||
| GenBank mRNA | JU319852.1 | - | 1685 | 96 | ||
| JU473095.1 | - | 1909 | 96 | |||
| JV043872.1 | - | 1658 | 96 | |||
Set Membership: |
|