Search
Search

HSPA4L| Gene Symbol: HSPA4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808421.3 | XP_002808467.2 | 2246 | 102 | ||
| XM_028848763.1 | XP_028704596.1 | 1802 | 102 | |||
| GenBank mRNA | JU333449.1 | - | 2156 | 102 | ||
| JV645493.1 | - | 2196 | 102 | |||
| JV709727.1 | - | 2183 | 102 | |||
Set Membership: |
|