Search
Search

FBLN2| Gene Symbol: FBLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130195.2 | XP_014985681.2 | 2693 | 78 | ||
| XM_015130196.2 | XP_014985682.2 | 2552 | 78 | |||
| GenBank mRNA | JV054694.1 | - | 2352 | 78 | ||
| JV645183.1 | - | 2509 | 78 | |||
Set Membership: |
|