Search
Search

ALKBH1| Gene Symbol: ALKBH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851723.1 | XP_028707556.1 | 762 | 70 | ||
| XM_028851724.1 | XP_028707557.1 | 768 | 70 | |||
| GenBank mRNA | JU326015.1 | - | 691 | 70 | ||
| JU477664.1 | - | 751 | 70 | |||
| JV726078.1 | - | 741 | 70 | |||
Set Membership: |
|