Search
Search

MAPKAP1| Gene Symbol: MAPKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116824.2 | XP_014972310.1 | 839 | 77 | ||
| XM_015116825.2 | XP_014972311.1 | 829 | 77 | |||
| XM_015116826.2 | XP_014972312.1 | 839 | 77 | |||
| XM_015116829.2 | XP_014972315.1 | 839 | 77 | |||
| XM_015116831.2 | XP_014972317.1 | 501 | 77 | |||
| XM_015116832.2 | XP_014972318.1 | 422 | 77 | |||
| XM_028834833.1 | XP_028690666.1 | 839 | 77 | |||
| XM_028834834.1 | XP_028690667.1 | 839 | 77 | |||
| GenBank mRNA | JU336352.1 | - | 820 | 77 | ||
| JU336353.1 | - | 820 | 77 | |||
| JU471530.1 | - | 810 | 77 | |||
| JU471531.1 | - | 810 | 77 | |||
| JV048227.1 | - | 813 | 77 | |||
| JV048228.1 | - | 813 | 77 | |||
| JV723214.1 | - | 500 | 77 | |||
Set Membership: |
|