Search
Search

CNKSR2| Gene Symbol: CNKSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086429.4 | XP_001086429.1 | 2549 | 73 | ||
| XM_001086538.4 | XP_001086538.1 | 2507 | 73 | |||
| XM_002806163.3 | XP_002806209.1 | 2459 | 73 | |||
| XM_002806164.3 | XP_002806210.1 | 2402 | 73 | |||
| XM_015127077.2 | XP_014982563.1 | 2459 | 73 | |||
| XM_028842224.1 | XP_028698057.1 | 2402 | 73 | |||
| XM_028842225.1 | XP_028698058.1 | 2312 | 73 | |||
| XM_028842226.1 | XP_028698059.1 | 2312 | 73 | |||
| GenBank mRNA | JU324607.1 | - | 2252 | 73 | ||
| JU324608.1 | - | 2105 | 73 | |||
| JV600260.1 | - | 2248 | 73 | |||
| JV600261.1 | - | 2101 | 73 | |||
| JV709803.1 | - | 2326 | 73 | |||