Search
Search

LRRK2| Gene Symbol: LRRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151449.2 | XP_015006935.2 | 1768 | 76 | ||
| XM_015151450.2 | XP_015006936.2 | 1542 | 76 | |||
| XM_015151451.2 | XP_015006937.2 | 1772 | 76 | |||
| GenBank mRNA | JV706586.1 | - | 1405 | 76 | ||
Set Membership: |
|