Search
Search

HLTF| Gene Symbol: HLTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131773.2 | XP_014987259.2 | 1970 | 104 | ||
| XM_015131775.2 | XP_014987261.2 | 1970 | 104 | |||
| XM_015131776.2 | XP_014987262.2 | 1967 | 104 | |||
| XM_015131777.2 | XP_014987263.2 | 1970 | 104 | |||
| XM_015131778.2 | XP_014987264.2 | 1970 | 104 | |||
| XM_015131779.2 | XP_014987265.2 | 1967 | 104 | |||
| GenBank mRNA | JU321310.1 | - | 1961 | 104 | ||
| JU321312.1 | - | 1961 | 104 | |||
| JU470668.1 | - | 1954 | 104 | |||
| JV043207.1 | - | 1956 | 104 | |||
| JV043208.1 | - | 1956 | 104 | |||
| JV043209.1 | - | 1956 | 104 | |||
| JV047991.1 | - | 1956 | 104 | |||
Set Membership: |
|