Search
Search

CCDC88A| Gene Symbol: CCDC88A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112380.2 | XP_014967866.1 | 1678 | 132 | ||
| XM_015112381.2 | XP_014967867.1 | 1184 | 132 | |||
| XM_015112382.2 | XP_014967868.1 | 1184 | 132 | |||
| XM_015112384.2 | XP_014967870.1 | 1243 | 132 | |||
| XM_015112385.2 | XP_014967871.1 | 1184 | 132 | |||
| XM_015112386.2 | XP_014967872.1 | 1243 | 132 | |||
| XM_015112388.2 | XP_014967874.1 | 1184 | 132 | |||
| XM_015112389.2 | XP_014967875.1 | 1184 | 132 | |||
| XM_015112390.2 | XP_014967876.1 | 1184 | 132 | |||
| XM_015112391.2 | XP_014967877.1 | 1243 | 132 | |||
| GenBank mRNA | JU322214.1 | - | 1156 | 132 | ||
| JU335400.1 | - | 1156 | 132 | |||
| JU470692.1 | - | 1205 | 132 | |||
| JU476348.1 | - | 1205 | 132 | |||
Set Membership: |
|