Search
Search

COQ2| Gene Symbol: COQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808408.3 | XP_002808454.2 | 416 | 68 | ||
| GenBank mRNA | JU326296.1 | - | 118 | 68 | ||
| JV058094.1 | - | 219 | 68 | |||
| JV712184.1 | - | 235 | 68 | |||
Set Membership: |
|