Search
Search

MYO10| Gene Symbol: MYO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804314.3 | XP_002804360.2 | 4516 | 88 | ||
| XM_028849350.1 | XP_028705183.1 | 3966 | 88 | |||
| GenBank mRNA | JU327624.1 | - | 1563 | 88 | ||
| JV043423.1 | - | 4515 | 88 | |||
| JV043424.1 | - | 4515 | 88 | |||
| JV636823.1 | - | 4510 | 88 | |||
| JV715684.1 | - | 1568 | 88 | |||