Search
Search

DHX8| Gene Symbol: DHX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119814.2 | XP_014975300.1 | 1387 | 86 | ||
| XM_015119815.2 | XP_014975301.1 | 1387 | 86 | |||
| XM_028836540.1 | XP_028692373.1 | 1381 | 86 | |||
| GenBank mRNA | JU333202.1 | - | 1282 | 86 | ||
| JU474515.1 | - | 1300 | 86 | |||
| JV046324.1 | - | 1251 | 86 | |||
Set Membership: |
|