Search
Search

RAP1GAP2| Gene Symbol: RAP1GAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835878.1 | XP_028691711.1 | 555 | 60 | ||
| XM_028835879.1 | XP_028691712.1 | 510 | 60 | |||
| XM_028835880.1 | XP_028691713.1 | 584 | 60 | |||
| XM_028835881.1 | XP_028691714.1 | 589 | 60 | |||
| XM_028835882.1 | XP_028691715.1 | 549 | 60 | |||
| XM_028835883.1 | XP_028691716.1 | 756 | 60 | |||
| GenBank mRNA | JU333557.1 | - | 568 | 60 | ||
| JV567104.1 | - | 583 | 60 | |||
Set Membership: |
|