Search
Search

MAPKBP1| Gene Symbol: MAPKBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142007.2 | XP_014997493.2 | 1580 | 77 | ||
| XM_015142009.2 | XP_014997495.2 | 1562 | 77 | |||
| XM_015142010.2 | XP_014997496.2 | 1580 | 77 | |||
| XM_015142011.2 | XP_014997497.2 | 1562 | 77 | |||
| XM_028850770.1 | XP_028706603.1 | 1580 | 77 | |||
| XM_028850771.1 | XP_028706604.1 | 1562 | 77 | |||
| XM_028850772.1 | XP_028706605.1 | 1580 | 77 | |||
| XM_028850773.1 | XP_028706606.1 | 1580 | 77 | |||
| XM_028850774.1 | XP_028706607.1 | 1460 | 77 | |||
| GenBank mRNA | JU333509.1 | - | 1538 | 77 | ||
| JU333510.1 | - | 1556 | 77 | |||
| JU474778.1 | - | 1464 | 77 | |||
| JV044301.1 | - | 1537 | 77 | |||
| JV565851.1 | - | 1570 | 77 | |||