Search
Search

MAPKBP1| Gene Symbol: MAPKBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142007.2 | XP_014997493.2 | 3298 | 101 | ||
| XM_015142009.2 | XP_014997495.2 | 3280 | 101 | |||
| XM_015142010.2 | XP_014997496.2 | 3298 | 101 | |||
| XM_015142011.2 | XP_014997497.2 | 3280 | 101 | |||
| XM_028850770.1 | XP_028706603.1 | 3310 | 101 | |||
| XM_028850771.1 | XP_028706604.1 | 3292 | 101 | |||
| XM_028850772.1 | XP_028706605.1 | 3310 | 101 | |||
| XM_028850773.1 | XP_028706606.1 | 3310 | 101 | |||
| XM_028850774.1 | XP_028706607.1 | 3190 | 101 | |||
| GenBank mRNA | JU333509.1 | - | 3256 | 101 | ||
| JU333510.1 | - | 3274 | 101 | |||
| JU474778.1 | - | 3182 | 101 | |||
| JV044301.1 | - | 3255 | 101 | |||
| JV565851.1 | - | 3288 | 101 | |||
Set Membership: |
|