Search
Search

SLC25A33| Gene Symbol: SLC25A33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147599.1 | XP_015003085.1 | 283 | 63 | ||
| GenBank mRNA | JU336693.1 | - | 278 | 63 | ||
| JU473427.1 | - | 127 | 63 | |||
| JV045646.1 | - | 88 | 63 | |||
| JV630853.1 | - | 247 | 63 | |||
Set Membership: |
|