Search
Search

DLGAP2| Gene Symbol: DLGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852306.1 | XP_028708139.1 | 1171 | 88 | ||
| XM_028852307.1 | XP_028708140.1 | 871 | 88 | |||
| XM_028852308.1 | XP_028708141.1 | 1171 | 88 | |||
| XM_028852309.1 | XP_028708142.1 | 793 | 88 | |||
| XM_028852310.1 | XP_028708143.1 | 565 | 88 | |||
| XM_028852311.1 | XP_028708144.1 | 488 | 88 | |||
| GenBank mRNA | JV562932.1 | - | 326 | 88 | ||
| JV633447.1 | - | 336 | 88 | |||
Set Membership: |
|