Search
Search

FTO| Gene Symbol: FTO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126148.2 | XP_014981634.1 | 1335 | 66 | ||
| XM_028841592.1 | XP_028697425.1 | 1125 | 66 | |||
| GenBank mRNA | JU326512.1 | - | 1068 | 66 | ||
| JV628438.1 | - | 1178 | 66 | |||
Set Membership: |
|