Search
Search

ZNF638| Gene Symbol: ZNF638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112616.2 | XP_014968102.2 | 6107 | 73 | ||
| XM_015112617.2 | XP_014968103.2 | 6104 | 73 | |||
| XM_015112619.2 | XP_014968105.2 | 6002 | 73 | |||
| XM_015112620.2 | XP_014968106.2 | 6062 | 73 | |||
| XM_015112621.2 | XP_014968107.2 | 6063 | 73 | |||
| XM_015112622.2 | XP_014968108.2 | 6060 | 73 | |||
| XM_015112623.2 | XP_014968109.2 | 6057 | 73 | |||
| GenBank mRNA | JU334035.1 | - | 6045 | 73 | ||
| JU334036.1 | - | 5991 | 73 | |||
| JU475207.1 | - | 5991 | 73 | |||
| JV046781.1 | - | 5952 | 73 | |||
| JV541415.1 | - | 4441 | 73 | |||
| JV565102.1 | - | 6035 | 73 | |||