Search
Search

GRIK5| Gene Symbol: GRIK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838860.1 | XP_028694693.1 | 2289 | 52 | ||
| XM_028838861.1 | XP_028694694.1 | 2289 | 52 | |||
| XM_028838862.1 | XP_028694695.1 | 1790 | 52 | |||
| XM_028838863.1 | XP_028694696.1 | 1296 | 52 | |||
| XM_028838864.1 | XP_028694697.1 | 1444 | 52 | |||
| GenBank mRNA | JU334148.1 | - | 1305 | 52 | ||
| JV614285.1 | - | 1306 | 52 | |||
Set Membership: |
|