Search
Search

MAP3K5| Gene Symbol: MAP3K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002803926.3 | XP_002803972.2 | 2885 | 80 | ||
| XM_015137368.2 | XP_014992854.2 | 1481 | 80 | |||
| GenBank mRNA | JU327406.1 | - | 1192 | 80 | ||
| JV044647.1 | - | 1420 | 80 | |||
| JV047091.1 | - | 1420 | 80 | |||
| JV636163.1 | - | 1601 | 80 | |||
| JV715280.1 | - | 1190 | 80 | |||
Set Membership: |
|