Search
Search

ZNF782| Gene Symbol: ZNF782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118125.2 | XP_014973611.2 | 1054 | 129 | ||
| XM_015118126.2 | XP_014973612.2 | 858 | 129 | |||
| GenBank mRNA | JU481277.1 | - | 518 | 129 | ||
| JU481278.1 | - | 518 | 129 | |||
Set Membership: |
|