Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07228016_g1
See other USP19 GE Assays ›
Gene Symbol:
USP19
Gene Name:
ubiquitin specific peptidase 19
Gene Aliases:
-
Chromosome Location:
Chr.2: 105190992 - 105203837 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07228016_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: USP19
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015130929.2
XP_014986415.2
3532
85
XM_015130935.2
XP_014986421.2
3672
85
XM_015130936.2
XP_014986422.2
3538
85
XM_015130937.2
XP_014986423.2
3659
85
XM_015130941.2
XP_014986427.2
3495
85
XM_015130942.2
XP_014986428.1
3229
85
XM_015130944.2
XP_014986430.2
3430
85
XM_028844113.1
XP_028699946.1
3535
85
XM_028844114.1
XP_028699947.1
3665
85
XM_028844115.1
XP_028699948.1
3666
85
XM_028844116.1
XP_028699949.1
3538
85
XM_028844118.1
XP_028699951.1
3657
85
XM_028844119.1
XP_028699952.1
3529
85
XM_028844120.1
XP_028699953.1
3626
85
XM_028844121.1
XP_028699954.1
3499
85
XM_028844122.1
XP_028699955.1
3619
85
XM_028844123.1
XP_028699956.1
3493
85
XM_028844124.1
XP_028699957.1
3382
85
XM_028844125.1
XP_028699958.1
3241
85
XM_028844126.1
XP_028699959.1
3235
85
XM_028844127.1
XP_028699960.1
3235
85
XM_028844128.1
XP_028699961.1
3196
85
XM_028844129.1
XP_028699962.1
3190
85
GenBank mRNA
JV635919.1
-
3082
85
JV640704.1
-
3094
85
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Macaca mulatta
Panther Classification:
Molecular Function -
cysteine protease
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline