Search
Search

CHMP7| Gene Symbol: CHMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144987.2 | XP_015000473.1 | 1597 | 91 | ||
| XM_015144988.2 | XP_015000474.1 | 1565 | 91 | |||
| XM_028852464.1 | XP_028708297.1 | 1597 | 91 | |||
| XM_028852465.1 | XP_028708298.1 | 1597 | 91 | |||
| GenBank mRNA | JU320845.1 | - | 1004 | 91 | ||
| JU471576.1 | - | 1595 | 91 | |||
| JU471577.1 | - | 1563 | 91 | |||
| JV045610.1 | - | 1030 | 91 | |||
Set Membership: |
|