Search
Search

ATP8B4| Gene Symbol: ATP8B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850904.1 | XP_028706737.1 | 5660 | 103 | ||
| XM_028850905.1 | XP_028706738.1 | 5195 | 103 | |||
| XM_028850906.1 | XP_028706739.1 | 5568 | 103 | |||
| GenBank mRNA | JU348688.1 | - | 321 | 103 | ||
Set Membership: |
|