Search
Search

LRIG2| Gene Symbol: LRIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148525.2 | XP_015004011.2 | 1396 | 98 | ||
| XM_015148534.2 | XP_015004020.2 | 1396 | 98 | |||
| XM_015148540.2 | XP_015004026.2 | 1455 | 98 | |||
| GenBank mRNA | JU322799.1 | - | 1367 | 98 | ||
Set Membership: |
|