Search
Search

NUSAP1| Gene Symbol: NUSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141973.2 | XP_014997459.2 | 352 | 90 | ||
| XM_015141976.2 | XP_014997462.2 | 352 | 90 | |||
| XM_015141977.2 | XP_014997463.2 | 352 | 90 | |||
| XM_015141979.2 | XP_014997465.2 | 352 | 90 | |||
| XM_015141980.2 | XP_014997466.2 | 352 | 90 | |||
| XM_015141981.2 | XP_014997467.2 | 352 | 90 | |||
| XM_028850758.1 | XP_028706591.1 | 352 | 90 | |||
| XM_028850759.1 | XP_028706592.1 | 352 | 90 | |||
| XM_028850760.1 | XP_028706593.1 | 352 | 90 | |||
| XM_028850761.1 | XP_028706594.1 | 352 | 90 | |||
| XM_028850763.1 | XP_028706596.1 | 352 | 90 | |||
| GenBank mRNA | JU487739.1 | - | 286 | 90 | ||
| JV062720.1 | - | 170 | 90 | |||