Search
Search

NCAPD3| Gene Symbol: NCAPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799890.3 | XP_002799936.2 | 1800 | 113 | ||
| XM_028834182.1 | XP_028690015.1 | 1800 | 113 | |||
| GenBank mRNA | JU333638.1 | - | 1722 | 113 | ||
| JU472323.1 | - | 1716 | 113 | |||
| JV044614.1 | - | 1858 | 113 | |||
Set Membership: |
|