Search
Search

SMAP1| Gene Symbol: SMAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136674.2 | XP_014992160.2 | 841 | 61 | ||
| XM_015136675.2 | XP_014992161.2 | 744 | 61 | |||
| XM_015136676.2 | XP_014992162.2 | 627 | 61 | |||
| XM_015136680.2 | XP_014992166.2 | 772 | 61 | |||
| XM_028847444.1 | XP_028703277.1 | 643 | 61 | |||
| GenBank mRNA | JU335807.1 | - | 723 | 61 | ||
| JU335808.1 | - | 804 | 61 | |||
| JU335809.1 | - | 804 | 61 | |||
| JU476685.1 | - | 531 | 61 | |||
| JV047860.1 | - | 714 | 61 | |||
| JV489122.1 | - | 652 | 61 | |||
| JV720577.1 | - | 779 | 61 | |||
Set Membership: |
|