Search
Search

IQGAP2| Gene Symbol: IQGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140297.2 | XP_014995783.1 | 4795 | 99 | ||
| XM_015140299.2 | XP_014995785.1 | 4624 | 99 | |||
| XM_015140300.2 | XP_014995786.1 | 3103 | 99 | |||
| XM_015140301.2 | XP_014995787.1 | 3052 | 99 | |||
| XM_028849526.1 | XP_028705359.1 | 4557 | 99 | |||
| GenBank mRNA | JU474072.1 | - | 4711 | 99 | ||
Set Membership: |
|