Search
Search

NF1| Gene Symbol: NF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119003.2 | XP_014974489.1 | 6396 | 151 | ||
| XM_015119005.2 | XP_014974491.1 | 6387 | 151 | |||
| XM_015119007.2 | XP_014974493.1 | 6324 | 151 | |||
| XM_028836183.1 | XP_028692016.1 | 6333 | 151 | |||
| XM_028836184.1 | XP_028692017.1 | 6327 | 151 | |||
| GenBank mRNA | JU320262.1 | - | 6182 | 151 | ||
| JU320263.1 | - | 6119 | 151 | |||
Set Membership: |
|